Product Details

SNP ID
hCV26762409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:21467607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAAACCACATTCCATAAGAACC[A/G]CCGCTCATGGCCCTGTTCTTTTCAG
Phenotype
MIM: 612992
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NBPF3 PubMed Links

Gene Details

Gene
NBPF3
Gene Name
neuroblastoma breakpoint family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256416.2 Intron NP_001243345.1
NM_001256417.2 Intron NP_001243346.1
NM_032264.4 Intron NP_115640.1

View Full Product Details