Product Details

SNP ID
rs1863842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11307129 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATTAAAGTTAGAGCTATGATGA[A/C]CTTGTTCCAATGTCACGGCATTTGT
Phenotype
MIM: 180990
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRB4 PubMed Links

Gene Details

Gene
PRB4
Gene Name
proline rich protein BstNI subfamily 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261399.2 870 UTR 3 NP_001248328.1
NM_002723.5 870 UTR 3 NP_002714.2

View Full Product Details