Product Details

SNP ID
rs1863843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11307323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCTTAGGTTGCTGTGACTCATT[C/G]CTGTTGAATTTGTTCAGGGATATAT
Phenotype
MIM: 180990
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRB4 PubMed Links

Gene Details

Gene
PRB4
Gene Name
proline rich protein BstNI subfamily 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261399.2 Intron NP_001248328.1
NM_002723.5 Intron NP_002714.2

View Full Product Details