Product Details

SNP ID
rs7173655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74340021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTCATGTCACCAAAAACCCAC[C/T]CATGTCCACAGTGAGTAGAGTGTCA
Phenotype
MIM: 118485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC33 PubMed Links

Gene Details

Gene
CCDC33
Gene Name
coiled-coil domain containing 33
There are no transcripts associated with this gene.

Gene
CYP11A1
Gene Name
cytochrome P450 family 11 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000781.2 Intron NP_000772.2
NM_001099773.1 Intron NP_001093243.1

View Full Product Details