Product Details
- SNP ID
-
rs2629727
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:74211126 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCTAGGGGAAGTTTTGCCAGAGA[A/T]TCCCCCCCACCACTACGCACGCACA
- Phenotype
-
MIM: 610745
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36139986] are located under a probe and SNP(s) [rs141730608] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC33
- Gene Name
- coiled-coil domain containing 33
- Gene
- STRA6
- Gene Name
- stimulated by retinoic acid 6
View Full Product Details