Product Details

SNP ID
rs8086956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46193937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAGTGGATATTTACACACCTTTA[A/G]ATTAGCTCAAATGAAACTTTGAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C18orf25 PubMed Links

Gene Details

Gene
C18orf25
Gene Name
chromosome 18 open reading frame 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008239.2 Intron NP_001008240.1
NM_145055.3 Intron NP_659492.1
XM_011525822.1 Intron XP_011524124.1
XM_017025555.1 Intron XP_016881044.1

View Full Product Details