Product Details

SNP ID
rs3117578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31668254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCTTTGCACCTTCCAGTCTAGA[A/G]TTTCATCTTCTGCATTGACATTGTT
Phenotype
MIM: 142590 MIM: 115441 MIM: 142610 MIM: 610433 MIM: 610434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAG6 PubMed Links

Gene Details

Gene
BAG6
Gene Name
BCL2 associated athanogene 6
There are no transcripts associated with this gene.

Gene
C6orf47
Gene Name
chromosome 6 open reading frame 47
There are no transcripts associated with this gene.

Gene
CSNK2B
Gene Name
casein kinase 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282385.1 Intron NP_001269314.1
NM_001320.6 Intron NP_001311.3
Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
There are no transcripts associated with this gene.

Gene
LY6G5B
Gene Name
lymphocyte antigen 6 complex, locus G5B
There are no transcripts associated with this gene.

Gene
LY6G5C
Gene Name
lymphocyte antigen 6 complex, locus G5C
There are no transcripts associated with this gene.

View Full Product Details