Product Details

SNP ID
rs2523503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31545782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAGACCTAACATGCAACTTCAT[A/C]CTAAAACAGACCGTAATATCCCCAC
Phenotype
MIM: 606853 MIM: 142560 MIM: 601022
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1G2 PubMed Links

Gene Details

Gene
ATP6V1G2
Gene Name
ATPase H+ transporting V1 subunit G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204078.1 Intron NP_001191007.1
NM_130463.3 Intron NP_569730.1
NM_138282.2 Intron NP_612139.1
Gene
ATP6V1G2-DDX39B
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
DDX39B
Gene Name
DEAD-box helicase 39B
There are no transcripts associated with this gene.

Gene
DDX39B-AS1
Gene Name
DDX39B antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC100287329
Gene Name
uncharacterized LOC100287329
There are no transcripts associated with this gene.

Gene
NFKBIL1
Gene Name
NFKB inhibitor like 1
There are no transcripts associated with this gene.

Gene
SNORD117
Gene Name
small nucleolar RNA, C/D box 117
There are no transcripts associated with this gene.

Gene
SNORD84
Gene Name
small nucleolar RNA, C/D box 84
There are no transcripts associated with this gene.

View Full Product Details