Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195001.1 | Intron | NP_001181930.1 | |||
NM_005704.4 | Intron | NP_005695.3 | |||
NM_133177.3 | Intron | NP_573438.3 | |||
NM_133178.3 | Intron | NP_573439.2 | |||
XM_006710269.2 | Intron | XP_006710332.1 | |||
XM_016999992.1 | Intron | XP_016855481.1 | |||
XM_016999993.1 | Intron | XP_016855482.1 | |||
XM_016999994.1 | Intron | XP_016855483.1 | |||
XM_016999995.1 | Intron | XP_016855484.1 |