Product Details

SNP ID
rs3738067
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:28736724 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTGGGGGCGGGGAGGCCGTGAGCAC[A/G]GCTTCCGCTCCTCCAATCCGCCAGA
Phenotype
MIM: 610640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YTHDF2 PubMed Links

Gene Details

Gene
YTHDF2
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172828.1 101 Intron NP_001166299.1
NM_001173128.1 101 UTR 5 NP_001166599.1
NM_016258.2 101 UTR 5 NP_057342.2

View Full Product Details