Product Details

SNP ID
rs4327312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42206765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTTTCATTCTTTTACCATTTT[A/G]TTTATGTTTAAATAACTTCATTTCA
Phenotype
MIM: 603076
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG1 PubMed Links

Gene Details

Gene
ABCG1
Gene Name
ATP binding cassette subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004915.3 Intron NP_004906.3
NM_016818.2 Intron NP_058198.2
NM_207174.1 Intron NP_997057.1
NM_207627.1 Intron NP_997510.1
NM_207628.1 Intron NP_997511.1
NM_207629.1 Intron NP_997512.1
XM_011529806.1 Intron XP_011528108.1
XM_011529807.2 Intron XP_011528109.1

View Full Product Details