Product Details

SNP ID
rs2240379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:243585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGCTCGTGAAGGACTGTCAAT[A/G]TTGTCTTCAAGGTACTCTTCCCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF596 PubMed Links

Gene Details

Gene
ZNF596
Gene Name
zinc finger protein 596
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042415.2 Intron NP_001035880.1
NM_001042416.2 Intron NP_001035881.1
NM_001287254.1 Intron NP_001274183.1
NM_001287255.1 Intron NP_001274184.1
NM_001287256.1 Intron NP_001274185.1
NM_001287399.1 Intron NP_001274328.1
NM_173539.3 Intron NP_775810.2
XM_011534744.1 Intron XP_011533046.1
XM_017013166.1 Intron XP_016868655.1

View Full Product Details