Product Details

SNP ID
hCV26807466
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:111832414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGGTCTGTTCCAACAACTGGG[T/C]CCCAAATTTCAGTACCTTTAGCTCA
Phenotype
MIM: 608473
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
ANAPC1 PubMed Links

Gene Details

Gene
ANAPC1
Gene Name
anaphase promoting complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022662.3 Intron NP_073153.1

View Full Product Details