Product Details

SNP ID
rs35921651
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:153327675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTAGATTCAGTGGATGTCCTA[A/G]CAGTGACAATTCGGGGGTTTTTTTA
Phenotype
MIM: 608198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGLYRP4 PubMed Links

Gene Details

Gene
PGLYRP4
Gene Name
peptidoglycan recognition protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020393.3 Intron NP_065126.2
XM_011509789.2 Intron XP_011508091.1
XM_011509790.1 Intron XP_011508092.1
XM_011509791.2 Intron XP_011508093.1
XM_011509792.1 Intron XP_011508094.1
XM_011509793.2 Intron XP_011508095.1

View Full Product Details