Product Details
- SNP ID
-
rs4367782
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
50
- Location
-
Chr.1:111308092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTATTCATTCTATGACTCATAAAC[A/G]CAGTAATTCTTTCCAATAACCAAGT
- Phenotype
-
MIM: 606080
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHIA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80278879] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHIA
- Gene Name
- chitinase, acidic
View Full Product Details