Product Details

SNP ID
rs4803400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40900057 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCACCCTTCAGAACTTCTCTCTA[G/T]GCTCGCTGGTGCCACTTGTGGACAT
Phenotype
MIM: 601133
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2G1P PubMed Links

Gene Details

Gene
CYP2G1P
Gene Name
cytochrome P450 family 2 subfamily G member 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details