Product Details

SNP ID
rs2286804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46230998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGACCTGTCTGAGGCCCACCCTGC[A/C]GCTGCCCTGAGGAGGCCCACAGGTC
Phenotype
MIM: 610544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGFL1 PubMed Links

Gene Details

Gene
IGFL1
Gene Name
IGF like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198541.1 524 UTR 3 NP_940943.1

View Full Product Details