Product Details

SNP ID
rs11650073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39969627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTGCACAGAGTTAGGCGAGGTC[A/G]TTGAGACTGACAGGTAAGAGATCAT
Phenotype
MIM: 611218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMA PubMed Links

Gene Details

Gene
GSDMA
Gene Name
gasdermin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178171.4 213 Intron NP_835465.2
XM_006721832.2 213 Intron XP_006721895.1
XM_011524651.2 213 UTR 5 XP_011522953.1
XM_017024502.1 213 Intron XP_016879991.1
XM_017024503.1 213 Intron XP_016879992.1
XM_017024504.1 213 Intron XP_016879993.1

View Full Product Details