Product Details

SNP ID
rs2854843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45891536 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAAGTTATTCAAAATCTAGAAA[C/T]GTCTTCTGCTAAATTCTTAATTAAG
Phenotype
MIM: 146730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFBP1 PubMed Links

Gene Details

Gene
IGFBP1
Gene Name
insulin like growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000596.2 Intron NP_000587.1

View Full Product Details