Product Details

SNP ID
rs4407176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:362807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAGCTCCCCCATTCAAA[C/T]GGCCTGCTCATCAGCAGAGAAACGG
Phenotype
MIM: 609503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THEG PubMed Links

Gene Details

Gene
THEG
Gene Name
theg spermatid protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016585.4 Intron NP_057669.1
NM_199202.2 Intron NP_954672.1
XM_011528049.2 Intron XP_011526351.1
XM_011528050.2 Intron XP_011526352.1
XM_011528051.2 Intron XP_011526353.1
XM_011528052.2 Intron XP_011526354.1

View Full Product Details