Product Details

SNP ID
rs4361555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:144161967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCCTGTTCAATTTGTTACCAC[C/T]ATGACAATGGTAAGTAAACACAAAA
Phenotype
MIM: 613423 MIM: 611483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD16 PubMed Links

Gene Details

Gene
KCTD16
Gene Name
potassium channel tetramerization domain containing 16
There are no transcripts associated with this gene.

Gene
YIPF5
Gene Name
Yip1 domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024947.3 Intron NP_001020118.1
NM_001271732.1 Intron NP_001258661.1
NM_030799.8 Intron NP_110426.4

View Full Product Details