Product Details

SNP ID
rs13184323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:142115154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTATTAAGAAAGAAAAAAAG[G/T]CTCCAAGAGATTCAAGTGACATCCT
Phenotype
MIM: 612050
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NDFIP1 PubMed Links

Gene Details

Gene
NDFIP1
Gene Name
Nedd4 family interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030571.3 Intron NP_085048.1

View Full Product Details