Product Details

SNP ID
rs4041245
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54159155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTTCCTCCCTACATGACAAAGTCC[C/T]TTTCTTCTGCTGCTGCTCTTCTCTA
Phenotype
MIM: 603547
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MBD2 PubMed Links

Gene Details

Gene
MBD2
Gene Name
methyl-CpG binding domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003927.4 Intron NP_003918.1
NM_015832.4 Intron NP_056647.1

View Full Product Details