Product Details

SNP ID
rs12677998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38391683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGCTAGAGACCCTTAATTGGG[A/G]TGCCCAATTCTAATTTTTTTTTTTT
Phenotype
MIM: 607083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LETM2 PubMed Links

Gene Details

Gene
LETM2
Gene Name
leucine zipper and EF-hand containing transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199659.2 Intron NP_001186588.1
NM_001199660.2 Intron NP_001186589.1
NM_001286787.1 Intron NP_001273716.1
NM_001286819.1 Intron NP_001273748.1
NM_001286821.1 Intron NP_001273750.1
NM_144652.3 Intron NP_653253.1
XM_006716291.3 Intron XP_006716354.1
XM_011544406.1 Intron XP_011542708.1
XM_011544407.1 Intron XP_011542709.1
XM_011544408.2 Intron XP_011542710.1
XM_011544409.2 Intron XP_011542711.1
XM_011544410.2 Intron XP_011542712.1
XM_017013051.1 Intron XP_016868540.1
XM_017013052.1 Intron XP_016868541.1
XM_017013053.1 Intron XP_016868542.1
XM_017013054.1 Intron XP_016868543.1
XM_017013055.1 Intron XP_016868544.1
XM_017013056.1 Intron XP_016868545.1
XM_017013057.1 Intron XP_016868546.1
Gene
WHSC1L1
Gene Name
Wolf-Hirschhorn syndrome candidate 1-like 1
There are no transcripts associated with this gene.

View Full Product Details