Product Details

SNP ID
rs2600096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:5132600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGTTTCTAATAGTCTGTCCC[G/T]TTTCACAGAGCCAAGACTTTATCAT
Phenotype
MIM: 616596
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL8B PubMed Links

Gene Details

Gene
ARL8B
Gene Name
ADP ribosylation factor like GTPase 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018184.2 Intron NP_060654.1

View Full Product Details