Product Details

SNP ID
rs1802314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50325080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCAGACTCTGCCACGACATCAT[C/T]ATGGACTATTTACAAACCTTCACAT
Phenotype
MIM: 603551 MIM: 605082 MIM: 607052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYAL2 PubMed Links

Gene Details

Gene
HYAL2
Gene Name
hyaluronoglucosaminidase 2
There are no transcripts associated with this gene.

Gene
RASSF1
Gene Name
Ras association domain family member 1
There are no transcripts associated with this gene.

Gene
TUSC2
Gene Name
tumor suppressor candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007275.2 1513 UTR 3 NP_009206.1

View Full Product Details