Product Details
- SNP ID
-
rs13826
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:34562906 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTATGACGAGTGCACGATGATGGA[C/T]CACTGTCATGGGGAACACAGTGCGG
- Phenotype
-
MIM: 606098
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SNX6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113133395] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNX6
- Gene Name
- sorting nexin 6
View Full Product Details