Product Details
- SNP ID
-
rs4597303
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:88697153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTACCAAATGCGCAAGGTGGAAG[C/T]GGGGCCGGGGCAGGGTCGGGCTGGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTU2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117229196] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTU2
- Gene Name
- cytosolic thiouridylase subunit 2
There are no transcripts associated with this gene.
- Gene
- RNF166
- Gene Name
- ring finger protein 166
- Gene
- SNAI3-AS1
- Gene Name
- SNAI3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details