Product Details

SNP ID
rs34864824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46381226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCAGCCAGCAGGGCGCCAGGG[A/C]TGAGCCGGCCGTGGAGGGGAGCGGG
Phenotype
MIM: 118495 MIM: 601441 MIM: 162096
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHRM4 PubMed Links

Gene Details

Gene
CHRM4
Gene Name
cholinergic receptor muscarinic 4
There are no transcripts associated with this gene.

Gene
DGKZ
Gene Name
diacylglycerol kinase zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105540.1 285 Intron NP_001099010.1
NM_001199266.1 285 Intron NP_001186195.1
NM_001199267.1 285 Intron NP_001186196.1
NM_001199268.1 285 Intron NP_001186197.1
NM_003646.3 285 Intron NP_003637.2
NM_201532.2 285 Intron NP_963290.1
NM_201533.3 285 Intron NP_963291.2
XM_005253181.3 285 Intron XP_005253238.1
XM_005253182.3 285 Intron XP_005253239.1
XM_006718354.1 285 Intron XP_006718417.1
XM_006718355.2 285 Intron XP_006718418.1
XM_011520422.1 285 Intron XP_011518724.1
XM_011520423.1 285 Intron XP_011518725.1
XM_017018455.1 285 Intron XP_016873944.1
XM_017018456.1 285 Intron XP_016873945.1
XM_017018457.1 285 Intron XP_016873946.1
Gene
MDK
Gene Name
midkine (neurite growth-promoting factor 2)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012333.2 285 Intron NP_001012333.1
NM_001012334.2 285 UTR 5 NP_001012334.1
NM_001270550.1 285 Intron NP_001257479.1
NM_001270551.1 285 Intron NP_001257480.1
NM_001270552.1 285 Intron NP_001257481.1
NM_002391.4 285 Intron NP_002382.1
XM_011520116.2 285 UTR 5 XP_011518418.1
XM_017017764.1 285 Intron XP_016873253.1
Gene
MIR4688
Gene Name
microRNA 4688
There are no transcripts associated with this gene.

View Full Product Details