Product Details

SNP ID
rs12980170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1027287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGGCGGTAGCCGCTTTGCACG[G/T]TCTTTGTCTCCCATCTGACTAATTT
Phenotype
MIM: 602373 MIM: 611011
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNN2 PubMed Links

Gene Details

Gene
CNN2
Gene Name
calponin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303499.1 Intron NP_001290428.1
NM_001303501.1 Intron NP_001290430.1
NM_004368.3 Intron NP_004359.1
NM_201277.2 Intron NP_958434.1
Gene
RNU6-2
Gene Name
RNA, U6 small nuclear 2
There are no transcripts associated with this gene.

Gene
TMEM259
Gene Name
transmembrane protein 259
There are no transcripts associated with this gene.

View Full Product Details