Product Details

SNP ID
rs12407568
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:228647582 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCCCCTGCCCGCCCCTTCCCCC[G/T]GTTTATAAGGTTGCGACTGCGCCGG
Phenotype
MIM: 606366
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DUSP5P1 PubMed Links

Gene Details

Gene
DUSP5P1
Gene Name
dual specificity phosphatase 5 pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC105373132
Gene Name
uncharacterized LOC105373132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011544354.2 1299 UTR 5 XP_011542656.1
Gene
RHOU
Gene Name
ras homolog family member U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021205.5 1299 Intron NP_067028.1
Gene
RNA5S14
Gene Name
RNA, 5S ribosomal 14
There are no transcripts associated with this gene.

Gene
RNA5S15
Gene Name
RNA, 5S ribosomal 15
There are no transcripts associated with this gene.

Gene
RNA5S16
Gene Name
RNA, 5S ribosomal 16
There are no transcripts associated with this gene.

Gene
RNA5S17
Gene Name
RNA, 5S ribosomal 17
There are no transcripts associated with this gene.

View Full Product Details