Product Details

SNP ID
rs1136728
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:234605300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCACATAAGAGAGACCAGTCTGC[A/C]CTGAGTCATATATACTCCAACTTGA
Phenotype
MIM: 615332
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IRF2BP2 PubMed Links

Gene Details

Gene
IRF2BP2
Gene Name
interferon regulatory factor 2 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077397.1 3632 UTR 3 NP_001070865.1
NM_182972.2 3632 UTR 3 NP_892017.2

View Full Product Details