Product Details

SNP ID
rs10172824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218357400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGGCAGGAGTCTTCACCTGTC[C/T]GTGCCAGTGAGGAAGGCTGTGGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATIP PubMed Links

Gene Details

Gene
CATIP
Gene Name
ciliogenesis associated TTC17 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320865.1 182 Intron NP_001307794.1
NM_198559.1 182 Intron NP_940961.1
XM_005246539.4 182 Intron XP_005246596.1
XM_005246541.4 182 Intron XP_005246598.1
XM_011511148.2 182 Intron XP_011509450.1
XM_011511149.2 182 Intron XP_011509451.1
XM_011511150.2 182 Intron XP_011509452.1
XM_017004053.1 182 UTR 5 XP_016859542.1
Gene
CATIP-AS1
Gene Name
CATIP antisense RNA 1
There are no transcripts associated with this gene.

Gene
CATIP-AS2
Gene Name
CATIP antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details