Product Details

SNP ID
rs3019543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60983748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCGACAGAATTTATTCAGATTT[G/T]GCCAATGGTTCCAATAATATCTTCT
Phenotype
MIM: 186720
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD6 PubMed Links

Gene Details

Gene
CD6
Gene Name
CD6 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254750.1 Intron NP_001241679.1
NM_001254751.1 Intron NP_001241680.1
NM_006725.4 Intron NP_006716.3
XM_006718738.1 Intron XP_006718801.1
XM_006718739.1 Intron XP_006718802.1
XM_006718740.1 Intron XP_006718803.1
XM_006718741.1 Intron XP_006718804.1
XM_011545360.1 Intron XP_011543662.1
XM_011545362.1 Intron XP_011543664.1

View Full Product Details