Product Details

SNP ID
rs2959886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63763703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCATCAGGTACTCGCCCCGCCA[G/C]CGCGGCTGGTAGTTCCGCCGCTGCT
Phenotype
MIM: 615699 MIM: 604249
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf95 PubMed Links

Gene Details

Gene
C11orf95
Gene Name
chromosome 11 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144936.1 1771 Silent Mutation CGC,CGG R584R NP_001138408.1
Gene
RTN3
Gene Name
reticulon 3
There are no transcripts associated with this gene.

View Full Product Details