Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001128612.2 | Intron | NP_001122084.1 | |||
NM_001128613.2 | Intron | NP_001122085.1 | |||
NM_001271831.1 | Intron | NP_001258760.1 | |||
NM_032344.3 | Intron | NP_115720.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033678.3 | Intron | NP_001028850.2 | |||
NM_001160389.1 | Intron | NP_001153861.1 | |||
NM_001160390.1 | Intron | NP_001153862.1 | |||
NM_001160392.1 | Intron | NP_001153864.1 | |||
NM_001160393.1 | Intron | NP_001153865.1 | |||
NM_031472.3 | Intron | NP_113660.1 | |||
XM_005274345.4 | Intron | XP_005274402.1 | |||
XM_005274346.4 | Intron | XP_005274403.1 | |||
XM_005274347.4 | Intron | XP_005274404.1 | |||
XM_006718706.3 | Intron | XP_006718769.1 | |||
XM_006718707.3 | Intron | XP_006718770.1 | |||
XM_017018400.1 | Intron | XP_016873889.1 | |||
XM_017018401.1 | Intron | XP_016873890.1 |