Product Details
- SNP ID
-
rs34383680
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:67274463 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATTGCTTTGCCTGTCATCTGCAT[C/T]TGCCTGTCGCTACCTTCCGCTTGAC
- Phenotype
-
MIM: 109635
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79631407] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRK2
- Gene Name
- G protein-coupled receptor kinase 2
View Full Product Details