Product Details

SNP ID
rs4466868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72097717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCACATTTTTCTCTGTGATTA[C/G]TAGCAGGTTAGGGTACTGTATAAGC
Phenotype
MIM: 614717 MIM: 613510 MIM: 612414
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANAPC15 PubMed Links

Gene Details

Gene
ANAPC15
Gene Name
anaphase promoting complex subunit 15
There are no transcripts associated with this gene.

Gene
LAMTOR1
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017907.2 754 UTR 3 NP_060377.1
Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 754 Intron NP_001138779.1
NM_001145308.4 754 Intron NP_001138780.1
NM_001145309.3 754 Intron NP_001138781.1
NM_001145310.3 754 Intron NP_001138782.1
NM_001205138.3 754 Intron NP_001192067.1
NM_001271471.2 754 Intron NP_001258400.1
NM_001318803.1 754 Intron NP_001305732.1
NM_145309.5 754 Intron NP_660352.1
XM_006718473.3 754 Intron XP_006718536.1
XM_006718474.3 754 Intron XP_006718537.1
XM_011544847.2 754 Intron XP_011543149.1
XM_011544848.2 754 Intron XP_011543150.1
XM_017017356.1 754 Intron XP_016872845.1
XM_017017357.1 754 Intron XP_016872846.1
XM_017017358.1 754 Intron XP_016872847.1
XM_017017359.1 754 Intron XP_016872848.1
XM_017017360.1 754 Intron XP_016872849.1

View Full Product Details