Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199505.1 | Intron | NP_001186434.1 | |||
NM_001199506.1 | Intron | NP_001186435.1 | |||
NM_001281507.1 | Intron | NP_001268436.1 | |||
NM_080672.4 | Intron | NP_542403.1 | |||
NM_183244.1 | Intron | NP_899067.1 | |||
NM_183246.1 | Intron | NP_899069.1 | |||
XM_011528525.2 | Intron | XP_011526827.1 | |||
XM_011528526.2 | Intron | XP_011526828.1 | |||
XM_017027626.1 | Intron | XP_016883115.1 | |||
XM_017027627.1 | Intron | XP_016883116.1 | |||
XM_017027628.1 | Intron | XP_016883117.1 | |||
XM_017027629.1 | Intron | XP_016883118.1 | |||
XM_017027630.1 | Intron | XP_016883119.1 | |||
XM_017027631.1 | Intron | XP_016883120.1 |