Product Details

SNP ID
rs4658628
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:244352788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTCTCTCTCGCCTCATTTAGAGG[C/T]TGCATTACTAAATGGTTGACTGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf100 PubMed Links

Gene Details

Gene
C1orf100
Gene Name
chromosome 1 open reading frame 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012970.2 45 Intron NP_001012988.1
NM_001276348.1 45 Intron NP_001263277.1
NM_001276349.1 45 Intron NP_001263278.1
XM_011544124.2 45 Intron XP_011542426.1
XM_011544125.2 45 Intron XP_011542427.1
XM_011544128.2 45 UTR 5 XP_011542430.1
XM_011544129.2 45 Intron XP_011542431.1
XM_011544130.2 45 Intron XP_011542432.1
XM_017000571.1 45 Intron XP_016856060.1
XM_017000572.1 45 UTR 5 XP_016856061.1

View Full Product Details