Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012970.2 | 761 | Intron | NP_001012988.1 | ||
NM_001276348.1 | 761 | Intron | NP_001263277.1 | ||
NM_001276349.1 | 761 | Intron | NP_001263278.1 | ||
XM_011544124.2 | 761 | Intron | XP_011542426.1 | ||
XM_011544125.2 | 761 | Intron | XP_011542427.1 | ||
XM_011544128.2 | 761 | UTR 5 | XP_011542430.1 | ||
XM_011544129.2 | 761 | Intron | XP_011542431.1 | ||
XM_011544130.2 | 761 | Intron | XP_011542432.1 | ||
XM_017000571.1 | 761 | Intron | XP_016856060.1 | ||
XM_017000572.1 | 761 | UTR 5 | XP_016856061.1 |