Product Details

SNP ID
rs35650616
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:246728572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCACCTCCGGCAGCTCTGCTCT[A/G]TTTCCCTCAGGAATTCTCTCTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 Intron NP_057086.2

View Full Product Details