Product Details

SNP ID
rs4576684
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:247918972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGTCGTGAAACATTCCATTAAA[A/G]TGATACTTAAACATACAGCTGAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T8 PubMed Links

Gene Details

Gene
OR2T8
Gene Name
olfactory receptor family 2 subfamily T member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005522.1 Intron NP_001005522.1
XM_011544178.2 Intron XP_011542480.1
XM_017001172.1 Intron XP_016856661.1

View Full Product Details