Product Details

SNP ID
rs2713936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:56253787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAGGTGGAAAAAGTGGCTAAAA[A/C]TTACAGCTCTTCACAATCAGTGTCT
Phenotype
MIM: 612660
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RFX7 PubMed Links

Gene Details

Gene
RFX7
Gene Name
regulatory factor X7
There are no transcripts associated with this gene.

Gene
TEX9
Gene Name
testis expressed 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286449.1 Intron NP_001273378.1
NM_198524.2 Intron NP_940926.1
XM_005254359.4 Intron XP_005254416.1
XM_005254361.3 Intron XP_005254418.1
XM_011521530.2 Intron XP_011519832.1
XM_017022160.1 Intron XP_016877649.1
XM_017022161.1 Intron XP_016877650.1
XM_017022162.1 Intron XP_016877651.1
XM_017022163.1 Intron XP_016877652.1
XM_017022164.1 Intron XP_016877653.1
XM_017022166.1 Intron XP_016877655.1
XM_017022167.1 Intron XP_016877656.1

View Full Product Details