Product Details
- SNP ID
-
rs2469151
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:66706272 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGTGGCTCTAGGAATATTCTCCC[C/T]ATCTAAATCTGGACTAAGCCTTGGG
- Phenotype
-
MIM: 602931
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SMAD6
PubMed Links
Gene Details
- Gene
- SMAD6
- Gene Name
- SMAD family member 6
View Full Product Details