Product Details
- SNP ID
-
rs7434051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139019261 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAACACAATGCATGCTGAAAACAT[C/G]TGATAAAATTCAACACAAATTTTCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23B
PubMed Links
Gene Details
- Gene
- PRR23B
- Gene Name
- proline rich 23B
View Full Product Details