Product Details

SNP ID
rs4420644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9463304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGCTTCTGAAGCTAGGCAATTT[C/T]TAGGCTGTGAGACTAGGGTATCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105372269 PubMed Links

Gene Details

Gene
LOC105372269
Gene Name
endogenous retrovirus group K member 8 Gag polyprotein-like
There are no transcripts associated with this gene.

Gene
ZNF560
Gene Name
zinc finger protein 560
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152476.2 Intron NP_689689.2

View Full Product Details