Product Details

SNP ID
rs6965294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1819262 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACACGTTGGCTGTGGCTGGAAATA[C/T]TTCTGAGGCTGCCTGCGCGCAGGCT
Phenotype
MIM: 602686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAD1L1 PubMed Links

Gene Details

Gene
MAD1L1
Gene Name
MAD1 mitotic arrest deficient like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013836.1 Intron NP_001013858.1
NM_001013837.1 Intron NP_001013859.1
NM_001304523.1 Intron NP_001291452.1
NM_001304524.1 Intron NP_001291453.1
NM_001304525.1 Intron NP_001291454.1
NM_003550.2 Intron NP_003541.2
XM_005249877.1 Intron XP_005249934.1
XM_011515567.1 Intron XP_011513869.1
XM_011515568.2 Intron XP_011513870.1
XM_011515569.2 Intron XP_011513871.2
XM_011515571.1 Intron XP_011513873.1
XM_017012690.1 Intron XP_016868179.1
XM_017012691.1 Intron XP_016868180.1

View Full Product Details