Product Details

SNP ID
rs4549856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:115896674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGAAACATATTGATTAATGTT[C/G]TTTTAACTTAAGGAATGCAAAGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00474 PubMed Links

Gene Details

Gene
LINC00474
Gene Name
long intergenic non-protein coding RNA 474
There are no transcripts associated with this gene.

View Full Product Details