Product Details

SNP ID
rs4077098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137314356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTGCCAAAAGAACCTGGCCAG[G/T]CCCGGCACTGACTTTCCACTGGAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXD3 PubMed Links

Gene Details

Gene
EXD3
Gene Name
exonuclease 3'-5' domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286823.1 Intron NP_001273752.1
NM_017820.4 Intron NP_060290.3
XM_011518810.1 Intron XP_011517112.1
XM_011518814.1 Intron XP_011517116.1
XM_011518815.1 Intron XP_011517117.1
XM_017014861.1 Intron XP_016870350.1
XM_017014862.1 Intron XP_016870351.1
XM_017014863.1 Intron XP_016870352.1
XM_017014864.1 Intron XP_016870353.1
XM_017014865.1 Intron XP_016870354.1
XM_017014866.1 Intron XP_016870355.1
XM_017014867.1 Intron XP_016870356.1

View Full Product Details